missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit and Quant-it Assay Kit Standards

Product Code. p-7235450
Click to view available options
Quantity:
1 set
2 x 5 mL
5 each
5 mL
Product Type:
1X dsDNA HS Assay Lambda Std
Qubit 1X dsDNA BR Lambda Std
Qubit RNA IQ Standards
Qubit RNA XR Standard
Unit Size:
1 set
Each
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 15880210

Brand: Invitrogen™ Q33233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards.

Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards. The Qubit standards can be used in conjunction with different Qubit or Quant-iT nucleic acid quantitation assay kits and offer stand-alone versions of the kit standard for performing additional assays.

Qubit 1X dsDNA HS Assay Lambda Standard

Qubit 1X dsDNA HS Assay Lambda Standard can be used with either Qubit 1X dsDNA HS Assay Kits, Qubit dsDNA HS Assay Kits, or their corresponding Quant-iT Assay Kits. The standards are supplied at the same concentration as the highest concentration dsDNA standard (10 ng/μL) in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit 1X dsDNA BR Standards

Qubit 1X dsDNA BR (Broad Range) Standards can be used with either Qubit 1X dsDNA BR Assay Kits, Qubit dsDNA BR Assay Kits, or their corresponding Quant-iT Assay Kits. The Qubit 1X dsDNA BR Standards includes standards at two concentrations, 0 and 100 ng/μL. The high standard corresponds to the same concentration as the highest concentration dsDNA standard in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit RNA IQ Assay Standards

Qubit RNA IQ Assay RNA standards are designed for use with the Qubit RNA IQ Assay Kit. Sufficient amounts of the small and large and/or structured RNA standard are supplied for generating an IQ standard curve for up to 500 samples. The RNA IQ standards are designed to measure RNA degradation and are not interchangeable with the standards from other RNA quantitation assays, including the Qubit RNA HS, BR, or XR assay kits.

Note: Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex fluorometer.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Qubit™ dsDNA HS Assay – Lambda DNA Standard, 5 mL at 10 ng/μL in TE buffer
Store at 2°C to 8°C
Concentration 10 ng/μL
For Use With (Application) Qubit and Quant‐iT dsDNA HS Assays and 1 x dsDNA Assays
For Use With (Equipment) Qubit Fluorometer or fluorescence microplate reader
Green Features Fewer resources used, Less waste
Product Line Qubit
Product Type 1X dsDNA HS Assay Lambda Std
Quantity 5 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.