missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ dsDNA Quantification Assay Kits
GREENER_CHOICE

Product Code. 10616763
Click to view available options
Quantity:
100 assays
500 assays
Assay:
dsDNA Quantitation, BR
dsDNA Quantitation, HS
Quantitation Range:
0.1 to 120 ng
4 to 2000 ng
Unit Size:
1 set
100 assays
500 assays
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10616763

Brand: Invitrogen™ Q32854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

HS dsDNA Assay Kit

Achieve accurate and precise quantification of double-stranded DNA with Qubit dsDNA HS and BR Assay Kits. These dsDNA quantification kits enable quick and selective detection of low and high abundance DNA samples, and can distinguish dsDNA from ssDNA, RNA, protein, and free nucleotides.

Achieve accurate and precise quantification of dsDNA with Qubit dsDNA BR (Broad Range) Assay Kits. These dsDNA quantification kits enable quick and selective detection of low and high abundance DNA samples, and can distinguish dsDNA from ssDNA, RNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.

Qubit dsDNA HS and BR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA), RNA, protein, and free nucleotides. All kits provide concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a Qubit Fluorometer.

Qubit dsDNA HS Assay Kit

The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.005 to 120 ng/μL, providing a detection range of 0.1-120 ng.

Qubit dsDNA BR Assay Kit

The Qubit dsDNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 2,000 ng/μL, providing a detection range of 4-2,000 ng.

Notes

  • Qubit dsDNA HS and BR Assay kits can be used with any Qubit Fluorometer
  • Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
TRUSTED_SUSTAINABILITY

Specifications

Assay dsDNA Quantitation, HS
Detection Method Fluorescence
Excitation/Emission 510/527
For Use With (Equipment) Qubit Fluorometer
No. of Reactions 500 Reactions
Quantitation Range 0.1 to 120 ng
Product Line Qubit
Quantity 500 assays
Shipping Condition Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.