Learn More
Invitrogen™ Qubit™ Flex Assay Tubes
Strip of eight tubes designed for the Qubit™ Flex.
Brand:  Invitrogen™ Q33252
184.74 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Product Code. 16438283
  • £197.00 / Each
Estimated Shipment: 02-12-2024
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Capacity: 200 μL
  • Thin-walled
  • Polypropylene tubes
  • For use with the Qubit™ Flex Fluorometer
  • Quantity: 125 tube strips
  • Strips of eight tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
125 tube strips
Qubit Flex Fluorometer
DNA, RNA or Protein Quantitation