Learn More
Invitrogen™ Qubit™ Fluorometer International Power Supply
The Qubit Fluorometer International Power Supply is designed for use with either the Qubit 3 or Qubit 4 Fluorometer.
Brand:  Invitrogen™ A36204
Product Code. 15940493
  • £67.75 / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Qubit Fluorometer International Power Supply is designed for use with either the Qubit 3 or Qubit 4 Fluorometer. This power supply should not be used with the original or Qubit 2.0 Fluorometer. The product includes a power cord and four interchangeable adaptors and enables the Qubit to be plugged into outlets around the world.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Qubit™
Power Supply
Qubit 3 Fluorometer, Qubit 4 Fluorometer
Room Temperature
1 each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Qubit™ Fluorometer International Power Supply > Qubit™