Learn More
Invitrogen™ Qubit™ Fluorometer USB Flash Drive
The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer.
Brand:  Invitrogen™ Q46009
34.37 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 15843764
  • £42.43 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer. Only the Qubit 2.0, 3, and 4 fluorometers have an available USB port. Data exported from the Qubit Fluorometer are stored in CSV format and can be opened by programs such as Microsoft Excel.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Qubit™
USB Flash Drive
Qubit Fluorometer
Room Temperature
1 each