Learn More
Invitrogen™ Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Description
Requires
Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes
Achieve accurate and precise quantification of RNA with Qubit RNA HS, BR, and XR Assay Kits. These RNA quantification kits enable quick and selective detection of low and high abundance RNA samples, and can distinguish RNA from DNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.
Qubit RNA HS Assay Kit
The Qubit RNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.2 to 200 ng/μL, providing a detection range of 4−200 ng.
Qubit RNA BR Assay Kit
The Qubit RNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.5 to 1,200 ng/μL, providing a detection range of 10−1,200 ng.
Qubit RNA XR Assay Kit
The Qubit RNA XR (Extended Range) Assay Kit, designed for use with Qubit 4 and Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of high-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 5 to 20,000 ng/μL, providing a detection range of 100−20,000 ng.
Notes
• Qubit RNA HS and BR Assay kits can be used with any Qubit Fluorometer
• Qubit RNA XR Assay Kits are designed to be used only with Qubit 4 or Qubit Flex Fluorometers
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Assay | RNA Quantification, HS |
| Detection Method | Fluorescence |
| Excitation/Emission | 644/673 |
| For Use With (Equipment) | Qubit Fluorometer |
| No. of Reactions | 100 Reactions |
| Quantitation Range | 4 to 200 ng |
| Product Line | Quant-iT, Qubit |
| Quantity | 100 assays |
| Shipping Condition | Room Temperature |
Documents
Scientific Resources
Application Notes
Flyers
Brochures
Posters
Product Information
Manuals
Your input is important to us. Please complete this form to provide feedback related to the content on this product.