missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits

Product Code. 15840210
Click to view available options
Quantity:
100 assays
500 assays
Assay:
RNA Quantification, BR
RNA Quantification, HS
RNA Quantification, ext range
Unit Size:
1 set
500 assays
Each
Pack of 100
Pack of 500
This item is not returnable. View return policy

Product Code. 15840210

Brand: Invitrogen™ Q33223

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kits enable quick and sensitive detection of low and high abundance RNA, and can distinguish between RNA and DNA, protein, and free nucleotides.

Achieve accurate and precise quantification of RNA with Qubit RNA HS, BR, and XR Assay Kits. These RNA quantification kits enable quick and selective detection of low and high abundance RNA samples, and can distinguish RNA from DNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.

Qubit RNA HS, BR, and XR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for RNA over DNA, protein, and free nucleotides. All kits provide concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a Qubit Fluorometer.

Qubit RNA HS Assay Kit
The Qubit RNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.2 to 200 ng/μL, providing a detection range of 4−200 ng.

Qubit RNA BR Assay Kit
The Qubit RNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.5 to 1,200 ng/μL, providing a detection range of 10−1,200 ng.

Qubit RNA XR Assay Kit

The Qubit RNA XR (Extended Range) Assay Kit, designed for use with Qubit 4 and Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of high-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 5 to 20,000 ng/μL, providing a detection range of 100−20,000 ng.

Notes
• Qubit RNA HS and BR Assay kits can be used with any Qubit Fluorometer
• Qubit RNA XR Assay Kits are designed to be used only with Qubit 4 or Qubit Flex Fluorometers
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

TRUSTED_SUSTAINABILITY

Specifications

Assay RNA Quantification, ext range
Detection Method Fluorescence
For Use With (Equipment) Qubit 4 and Qubit Flex Fluorometer
No. of Reactions 100 Reactions
Quantitation Range 0.1 to 20 μg
Product Line Qubit
Quantity 100 assays
Shipping Condition Dry Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.