missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.

Brand:  Thermo Scientific™ S1-15-AOAC-CH-KIT

 View more versions of this product

Product Code. 17339832

  • £166.00 / Pack of 50
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Extraction Kits

Extract analytes of interest from a homogenized sample using extraction salts. Use these kits with 10 g or 15 g samples. Choose from:

  • Extraction Salts in Pouches, No Tubes
  • Extraction Salts in Pouches with 50 mL Tubes
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QuEChERS AOAC Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaOAc in Pouches; Ceramic Homogenizers; Empty 50 mL Tubes; 50/Pk.
Salts in Pouches, Tubes, Ceramic Homogenizers
6 g MgSO4, 1.5 g NaOAc
50 mL
QuEChERS Extraction Kit
QuEChERS sample preparation for AOAC 2007.01 Method
15 g sample
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.