missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS EN 15662 Method Clean-up Kits

Product Code. 17339852 Shop All Thermo Scientific Products
Click to view available options
Sample Type:
Fruits and Vegetables with Fats and Waxes
General Fruits and Vegetables
Highly Pigmented Fruits and Vegetables
Pigmented Fruits and Vegetables
Tube Size:
2 mL
15 mL
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 17339852

Brand: Thermo Scientific™ S215PENKIT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use Thermo Scientific™ QuEChERS EN 15662 Method Clean-up Kits to analyze complex sample matrices for residues, toxins, and other contaminants.

Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the EN 15662 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS EN 15662 Method Clean-Up Kits make sample preparation straight-forward and efficient, enabling you to confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, highly pigmented fruits and vegetables, and fruits and vegetables containing fats and waxes.

What is the purpose of the chemicals used in the EN 15662 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer; removes remaining water from sample
  • NaCl: induces phase separation between water content in sample and acetonitrile layer; removes residual water from sample
  • Trisodium Citrate: buffers the sample to stabilize pH
  • Disodium Citrate Sesquihydrate: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: (graphitized carbon black) removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY

Specifications

Description QuEChERS EN 15662 Method Clean-up Kit; 900 mg MgSO4, 150 mg PSA, 15 mg GCB prefilled in 15 mL Tubes; 50/Pk.
Type QuEChERS Clean-up Kit
Includes Salts in Prefilled Tubes
Method QuEChERS sample preparation for EN 15662 Method
Salt Composition 900 mg MgSO4, 150 mg PSA, 15 mg GCB
Sample Type Pigmented Fruits and Vegetables
Tube Size 15 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.