missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS EN 15662 Method Extraction Kits

Use Thermo Scientific™ QuEChERS Extraction Kits for the EN 15662 Method to analyze complex sample matrices for residues, toxins, and other contaminants.

Brand:  Thermo Scientific™ S1-10-EN-POT

 View more versions of this product

Product Code. 17389822

  • £129.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the EN 15662 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS EN 15662 Method Extraction Kits make sample preparation straight-forward and efficient, enabling you to confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Three different extraction kit formats are available for 10 g samples.

  • Extraction Salts in Pouches, no Tubes
  • Extraction Salts in Pouches with 50 mL Tubes
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers

What is the purpose of the chemicals used in the EN 15662 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer; removes remaining water from sample
  • NaCl: induces phase separation between water content in sample and acetonitrile layer; removes residual water from sample
  • Trisodium Citrate: buffers the sample to stabilize pH
  • Disodium Citrate Sesquihydrate: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: (graphitized carbon black) removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QuEChERS EN 15662 Method Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl, 1 g Na-Citrate, 0.5 g Disodium Citrate Sesquihydrate in Pouches; No Tubes; 50/Pk.
Salts in Pouches
4 g MgSO4, 1 g NaCl, 1 g Na-Citrate, 0.5 g Disodium Citrate Sesquihydrate
QuEChERS Extraction Kit
QuEChERS sample preparation for EN 15662 Method
10 g sample
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.