missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuickBag™ UVM1 Broth

Product Code. 15795483
Click to view available options
Quantity:
2 x 4.5 L
3 x 2.7 L
Unit Size:
Pack of 2
Pack of 3
This item is not returnable. View return policy

Product Code. 15795483

Brand: Thermo Scientific™ DQ0863A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Shorten your workflow with easy-to-prepare 2.7L or 4.5L media bags: simply merge the segments to reconstitute in two minutes.

Save time in preparing selective enrichment medium for Listeria monocytogenes, while conforming to the USDA-FSIS method, using Thermo Scientific™ QuickBag™ UVM1 Broth. The irradiated medium is supplied in transparent plastic bags—simply pop the seal to mix the water and medium segments to reconstitute in two minutes. QuickBag Enrichment Media are available in 2.7L and 4.5L sizes to suit your workflow. Medium can be dispensed through the outlet tube as required.

Shorten your workflow with FitBag UVM1 Broth. FitBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

  • Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description QuickBag UVM1 Broth
Format Convenience Format
Product Type Dehydrated Culture Media
Quantity 3 x 2.7 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.