Learn More
Quickfit™ Borosilicate Glass Air Condenser
Manufactured from Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Air Condenser creates condensation of materials with boiling points above 150°C.
Brand:  Quickfit C2/22
Product Code. 12518865
  • £44.40 / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Simple condensers for condensation of materials with boiling points above 150°C