Learn More
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
£160.00 - £322.00
Specifications
Material Borosilicate Glass
Type Allihn Condenser
For Use With (Application) Ideal for reflux work
View More Specs
Products 6
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
11338074
missing translation for 'viewDocuments'
Quickfit
CX7/02/SC
19/26
£160.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11348074
missing translation for 'viewDocuments'
Quickfit
CX7/03/SC
24/29
£161.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
12030073
missing translation for 'viewDocuments'
Quickfit
CX7/04/SC
29/32
£170.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11351845
missing translation for 'viewDocuments'
Quickfit
CX7/05/SC
34/35
£322.00
Each
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
11371845
missing translation for 'viewDocuments' Promotion Details
Quickfit
CX7/08/SC
50/42
£260.00
Each
In Stock
to see stock.
Edge
Add to basket
11361845
missing translation for 'viewDocuments'
Quickfit
CX7/06/SC
40/38
£189.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
Description
Description
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Specifications
Borosilicate Glass
Allihn Condenser
Ideal for reflux work
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Quickfit™ Allihn Condenser with Ground Cone