Learn More
Quickfit™ Borosilicate Glass Centrifuge Tubes
Centrifuge in sturdy glass tubes . QuickFit™ Borosilicate Glass Centrifuge Tubes come with a Quickfit™ socket to accept a tapered stopper.
Brand:  Quickfit BC24/C24R
 View more versions of this product
Product Code. 12969433
  • £399.00 / Pack of 10
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass with precision ground Quickfit™socket to accept tapered stopper
  • Conical or cylindrical shape
  • Each tube is marked with its weight to the nearest 0.1g to enable tubes of similar weight to be selected from any batch
  • Can be centrifuged at RCF values up to 3, 000
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 mL
24/20
115 mm
3000 x g
Borosilicate Glass
Cylindrical
28.5 mm