Learn More
Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head
Freezes volatile materials and vapors onto the cold surface. Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head prevents materials from damaging vacuum pumps.
Brand:  Pyrex™ MF28/52
117.76 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10536465
  • £301.00 / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Freezes volatile materials and vapors onto the cold surface. Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head prevents materials from damaging vacuum pumps.
  • Designed for use in vacuum systems
  • Prevents vapours from volatile materials from damaging pumps etc
  • Easy to use: - immerse trap into Dewar vessel containing refrigerant e.g., dry ice - volatile materials passing through system are frozen onto the cold surface
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cold trap head
Borosilicate Glass
VAcuum systems
34/35, 19/26
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head > Cone Size: 34/35 and 19/26; For use with: Vacuum systems