Learn More
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket
Supplied with detachable plastic screwthread connectors which allow 9mm flexible tubing to be fitted easily and safely. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket is a highly efficient condenser with a large cooling surface. Suitable for either reflux or distillation work.
Brand: Quickfit C3/13/SC
83.41 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Supplied with detachable plastic screwthread connectors which allow 9mm flexible tubing to be fitted easily and safely. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket is a highly efficient condenser with a large cooling surface. Suitable for either reflux or distillation work.
- Suitable for either reflux or distillation work
- Highly efficient condensers with a large cooling surface
- Supplied with detachable plastic screwthread connectors which allow 9mm flexible tubing to be fitted easily and safely
Specifications
Suitable for either reflux or distillation work | |
24/29 | |
Condenser Coil |
310 mm (Overall) 165 mm (Effective) | |
Borosilicate Glass |