Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Keep material securely dry. QuickFit™ Borosilicate glass drying angled cone to fit QuickFitT™ apparatus.
Brand: Quickfit MF23/1
31.63 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.05264kg
Description
- For use with a desiccant such as calcium chloride
- Ground glass cone to fit Quickfit™ apparatus
Specifications
Tube | |
14/23 | |
Borosilicate Glass | |
Quickfit™ apparatus |