Learn More
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
£74.70 - £302.00
Specifications
Design Type | Ground Socket |
---|---|
Material | Borosilicate Glass |
Quantity | 5 Pack |
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | For Use With | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | For Use With | Price | Quantity & Availability | |||
11352055
|
Quickfit
MF24/3/8 |
29 mm | 75 mL | Quickfit Hollow Stopper, Fisher Scientific product code 12508865 |
£230.00
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
10747645
|
Quickfit
MF24/1/6 |
19 mm | 19 mL | Quickfit Hollow Stopper, Fisher Scientific product code 10155824 |
£74.70
Pack of 5 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10715101
|
Quickfit
MF24/5 |
41 mm | 195 mL | Quickfit Hollow Stopper, Fisher Scientific product code 11349623 |
£302.00
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
12475316
|
Quickfit
MF24/0/4 |
14 mm | 5 mL | Quickfit Hollow Stopper, Fisher Scientific product code 12558855 |
£151.00
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
11382045
|
Quickfit
MF24/1 |
19 mm | 11 mL | Quickfit Hollow Stopper, Fisher Scientific product code 10155824 |
£74.70
Pack of 5 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
11392045
|
Quickfit
MF24/1/5 |
19 mm | 15 mL | Quickfit Hollow Stopper, Fisher Scientific product code 10155824 |
£160.00
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
11342055
|
Quickfit
MF24/3 |
29 mm | 54 mL | Quickfit Hollow Stopper, Fisher Scientific product code 12508865 |
£160.00
Pack of 5 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
10074474
|
Quickfit
MF24/2 |
23 mm | 19 mL | Quickfit Hollow Stopper, Fisher Scientific product code 12568855 |
£74.70
Pack of 5 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||
10497400
|
Quickfit
MF24/2/6 |
23 mm | 31 mL | Quickfit Hollow Stopper, Fisher Scientific product code 12568855 |
£160.00
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||
12569795
|
Quickfit
MF24/12/4 |
17 mm | 10 mL | Quickfit Stopper Cone, Fisher Scientific product code 15537960 |
£103.00
Pack of 5 |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||
Description
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
- Presicion-ground socket ensures exact fit for stopper
- Test tube sizes correspond with Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers
- Order tubes and stoppers separately
Specifications
Ground Socket | |
Borosilicate Glass | |
5 Pack |