missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Heavy Entrainer Receiver
Determine water or volitile oil content. The QuikFit™ borosilicate glass heavy entrainer receiver is perfect when the solvent entrainer is heavier than water.
Brand: Quickfit WE7/23/3
147.39 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Determine water or volitile oil content. The QuikFit™ borosilicate glass heavy entrainer receiver is perfect when the solvent entrainer is heavier than water.
- Complies with BS 756
- For the determination of water content in petroleum products using a solvent entrainer which is heavier than water
- Also suitable for the estimation of volatile oil content
- Highest quality borosilicate glass with graduated tube and Quickfit™ precision ground joints
Specifications
For the determination of water content in petroleum products using a solvent entrainer which is heavier than water | |
Borosilicate Glass | |
BS 756 | |
Heavy entrainer |
Heavy Entrainer Receiver | |
3mL | |
3mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction