Learn More
Quickfit™ Borosilicate Glass Iodine Flask
Complies with BS 2735
Brand: N/A FIB500/4
Code : NEW
Additional Details : Weight : 0.52000kg
Description
Complies with BS 2735
- Complies with BS 2735
- For the determination of iodine
- Flask has ground socket and cup shaped top
- Stopper has extended handle which projects well above the liquid seal
Note: Stopper is not included; sold separately.
Specifications
BS 2735 | |
500 mL | |
2 | |
Iodine stopper: SIB 29 |
Top style: Cup shaped | |
Borosilicate Glass | |
Iodine Flask | |
29/32 |