missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Jacketed Coil Condenser

Complies with DIN 12593. Quickfit™ Borosilicate Glass Jacketed Coil Condenser has an outer water jacket which provides a large cooling surface. Jacket avoids vapour creeping around the coil. Ideal for reflux work.

£276.00 - £514.00

Specifications

Certifications/Compliance DIN 12593
Material Borosilicate Glass
Type Jacketed Coil Condenser
For Use With (Application) Ideal for reflux work
Length (Metric) Jacket 160 mm
View More Specs

Products 4
Product Code Brand Standard Taper Joint No Description Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Description Price Quantity & Availability  
12010073
View Documents
Quickfit
CX6/33/SC
24/29 Overall Length: 365mm
£399.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
12000073
View Documents
Quickfit
CX6/22/SC
19/26 Overall Length: 360mm
£385.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
15587780
View Documents
Quickfit
CX6/05/SC
34/35 Overall Length: 380mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15597780
View Documents
Quickfit
CX6/44/SC
29/32 Overall Length: 370mm
£514.00
Pack of 2
Estimated Shipment: 23-07-2025
to see stock.
Add to basket
 
Description

Description

  • Complies with DIN 12593
  • Coil condenser has an outer water jacket
  • Coil provides large cooling surface
  • Jacket avoids vapour creeping around the coil
  • Ideal for reflux work
Specifications

Specifications

DIN 12593
Jacketed Coil Condenser
160 mm
Borosilicate Glass
Ideal for reflux work
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quickfit™ Borosilicate Glass Jacketed Coil Condenser

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.