Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Product Code. 12039652
Click to view available options
Length (Metric):
135 mm
260 mm
340 mm
380 mm
390 mm
400 mm
530 mm
535 mm
550 mm
Unit Size:
Each
Product Code. 12039652
Brand: Quickfit C1/13/SC
Only null left
Add to basket
Edge
Add to basket
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN 12576
Description Quickfit™ Cone Size: 24/29
For Use With (Application) Suitable for distillation and reflux applications
Length (Metric) 390 mm
Standard Taper Joint No 24/29
Material Borosilicate Glass
Type Liebig Condenser