Learn More
Quickfit™ Medium Neck Flat Bottom Flask
Useful in reducing problems associated with splashing and frothing.
Brand: Quickfit FF250/3M
44.80 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Made of Borosilicate glass
- With longer neck than standard Quickfit flasks
Specifications
250 mL | |
Borosilicate Glass | |
150 mm | |
Medium |