Learn More
Quickfit™ Borosilicate Glass Pear Shaped Claisen-Vigreux Flask
Incorporates short, Vigreux type fractionating column.
Brand: Quickfit MD4/100
This item is not returnable.
View return policy
Description
- Column indents aid intimate contact between vapor and liquid
Specifications
Pear | |
100 mL | |
Borosilicate Glass |