Learn More
Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
£72.00 - £810.42
Specifications
Certifications/Compliance | ISO 1773, ISO 4797 |
---|---|
Material | Borosilicate Glass |
Bottom Shape | Round |
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | ||||
10459330
|
Quickfit
FR1L/3S |
1000 mL | 24/29 |
£120.74
Pack of 2 |
In Stock
Log in to see stock. |
||||
10488560
|
Quickfit
FR1L/4S |
1000 mL | 29/32 |
£133.00
Pack of 2 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
10691821
|
Quickfit
FR1L/5S |
1000 mL | 34/35 |
£152.74
Pack of 2 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
10829580
|
Quickfit
FR100/1S |
100 mL | 14/23 |
£185.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10744711
|
Quickfit
FR100/2S |
100 mL | 19/26 |
£177.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10538190
|
Quickfit
FR100/3S |
100 mL | 24/29 |
£181.63
Pack of 5 |
In Stock
Log in to see stock. |
||||
10580491
|
Quickfit
FR100/5S |
100 mL | 34/35 |
£148.00
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
10660871
|
Quickfit
FR10/0S |
10 mL | 10/19 |
£72.00
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
10121150
|
Quickfit
FR10/1S |
10 mL | 14/23 |
£134.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10365470
|
Quickfit
FR150/2S |
150 mL | 19/26 |
£154.80
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
10671071
|
Quickfit
FR150/3S |
150 mL | 24/29 |
£211.56
Pack of 5 |
In Stock
Log in to see stock. |
||||
12570086
|
Quickfit
FR150/4S |
150 mL | 29/32 |
£155.00
Pack of 5 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
12458976
|
Quickfit
FR150/5S |
150 mL | 34/35 |
£154.00
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
12000663
|
Quickfit
FR2L/3S |
2000 mL | 24/29 |
£115.00
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
10820861
|
Quickfit
FR250/1S |
250 mL | 14/23 |
£98.80
Pack of 2 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||||
10765111
|
Quickfit
FR25/1S |
25 mL | 14/23 |
£122.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
11391955
|
Quickfit
FR250/5S |
250 mL | 34/35 |
£227.00
Pack of 5 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
10765301
|
Quickfit
FR25/2S |
25 mL | 19/26 |
£138.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10558590
|
Quickfit
FR25/3S |
25 mL | 24/29 |
£140.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
12682002
|
Quickfit
FR250/3S |
250 mL | 24/29 |
£200.21
Pack of 5 |
In Stock
Log in to see stock. |
||||
12070673
|
Quickfit
FR5L/4S |
5 L | 29/32 |
£219.82
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
11381965
|
Quickfit
FR5L/5US |
5 L | 34/35 |
£235.30
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
10395530
|
Quickfit
FR500/2S |
500 mL | 19/26 |
£245.00
Pack of 5 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
10488570
|
Quickfit
FR500/3S |
500 mL | 24/29 |
£236.33
Pack of 5 |
In Stock
Log in to see stock. |
||||
10417620
|
Quickfit
FR500/5S |
500 mL | 34/35 |
£254.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10809900
|
Quickfit
FR50/1S |
50 mL | 14/23 |
£144.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
10181780
|
Quickfit
FR50/3S |
50 mL | 24/29 |
£155.83
Pack of 5 |
In Stock
Log in to see stock. |
||||
10407430
|
Quickfit
FR5/1S |
5 mL | 14/23 |
£144.00
Pack of 5 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
15507860
|
Quickfit
FR1L/2S |
1000 mL | 19/26 |
£283.80
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15517880
|
Quickfit
FR2L/5S |
2000 mL | 34/35 |
£448.00
Pack of 5 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
15577880
|
Quickfit
FR3L/3S |
3000 mL | 24/29 |
£626.00
Pack of 5 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
15587880
|
Quickfit
FR3L/5S |
3000 mL | 34/35 |
£519.00
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
15557890
|
Quickfit
FR5/0S |
5 mL | 10/19 |
£289.00
Pack of 20 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
10568590
|
Quickfit
FR250/2S |
250 mL | 19/26 |
£211.00
Pack of 5 |
In Stock
Log in to see stock. |
||||
15517870
|
Quickfit
FR1L/7S |
1000 mL | 45/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15587850
|
Quickfit
FR10L/5S |
10 L | 34/35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15577890
|
Quickfit
FR50/5S |
50 mL | 34/35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15527900
|
Quickfit
FR6L/7S |
6 L | 45/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Descrizione
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
- Hgh quality Quickfit™ ground glass socket
- Comprehensive range of 46 flasks from 5mL to 10,000mL
- Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
Specifica
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
Round |