missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle

Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.

Brand:  Quickfit MF48/13/500

76.45 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11342065

  • £189.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.

  • Complies with BS 2461
  • Sintered dip tube disperses gas into bubbles for greater efficiency
  • Choice of sinter porosities to achieve fine bubble size
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BS 2461
Borosilicate Glass
Bottle Heads
Stem Length: 217 mm
For model MF29/3/500 Dreschel bottle
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.