Learn More
Quickfit™ Three-Vertical-Neck Round-Bottom Flask
Includes three necks for easy addition of measurement tools. The Quickfit™ Three-Vertical-Neck Round-Bottome Flask is made from borosilicate glass.
Brand:  Quickfit FR3L/5S/22P
718.87 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15507890
Description
Description
  • Fitted with three Quickfit™ precision-ground sockets - One vertical centre neck - Two angled side necks
  • Quickfit™ ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3000 mL
249 mm
34/35 (Centre), 19/26 (side)
Borosilicate Glass
3
Round