Learn More
Quickfit™ Three-Vertical-Neck Round-Bottom Flask
Includes three necks for easy addition of measurement tools. The Quickfit™ Three-Vertical-Neck Round-Bottome Flask is made from borosilicate glass.
Brand:  Quickfit FR2L/5S/22P
 View more versions of this product
Product Code. 15537880
Description
Description
  • Fitted with three precision-ground sockets
    • One vertical center neck
    • Two vertical side necks
  • Ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2000 mL
228 mm
34/35 (Centre), 19/26 (side)
Borosilicate Glass
3
Round