Learn More
Quickfit™ Three-Vertical-Neck Round-Bottom Flask
Includes three necks for easy addition of measurement tools. The Quickfit™ Three-Vertical-Neck Round-Bottome Flask is made from borosilicate glass.
Brand: Quickfit FR2L/7S/44P
This item is not returnable.
View return policy
Description
- Fitted with three precision-ground sockets
- One vertical center neck
- Two vertical side necks
- Ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
Specifications
2000 mL | |
233 mm | |
45/40 (Centre), 29/32 (side) |
Borosilicate Glass | |
3 | |
Round |