Learn More
Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask
Manufactured from borosilicate glass. Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask's wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.
Brand: Quickfit FR1LF
113.86 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Manufactured from borosilicate glass
- With large diameter Pyrex Labware™ Quickfit™ flat-flange joint
- For use with Pyrex Labware™ Quickfit™ flat-flange lids and retaining clips
- Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues
Specifications
1000 mL | |
For use with Quickfit™ flat-flange lids and retaining clips | |
Wide |
Borosilicate Glass | |
155 mm | |
Round |