Learn More
Quickfit™ Distillation Splash Head Adapter
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
Brand: Quickfit SH7/53
106.58 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode - Lower cone accepts flask, upper cone accepts condenser - Splash head reduces entrainment of raw liquid into the vapour streamSpecifications
Distillation Splash Head Adapter | |
Borosilicate Glass (3.3) | |
Distillation assemblies in which the condenser is assembled in the vertical mode | |
24/29 (Condenser), 24/29 (flask) |