Learn More
Quickfit™ Distillation Splash Head Adapter
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
£90.35 - £267.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Distillation assemblies in which the condenser is assembled in the vertical mode |
Product Type | Distillation Splash Head Adapter |
Product Code | Brand | Cone Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Price | Quantity & Availability | ||||||
12078493
|
Quickfit
SH7/12 |
19/26 (Condenser), 19/26 (flask) |
£110.00
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
12415396
|
Quickfit
SH7/13 |
19/26 (Condenser), 24/29 (flask) |
£90.35
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
12435396
|
Quickfit
SH7/53 |
24/29 (Condenser), 24/29 (flask) |
£267.00
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
Description
Recieve distillate from a condenser. The QuickFit™ distillation splash head reduces the risk of raw liquid entering into the vapor stream.
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode - Lower cone accepts flask, upper cone accepts condenser - Splash head reduces entrainment of raw liquid into the vapour streamSpecifications
Borosilicate Glass (3.3) | |
Distillation assemblies in which the condenser is assembled in the vertical mode | |
Distillation Splash Head Adapter |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Quickfit™ Distillation Splash Head Adapter