Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand: Quickfit XA53
70.21 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
- Useful for the connection of dissimilar sized ground glass joints
- Large sized socket at the top and small sized cone at the base
Specifications
24/29 | |
Borosilicate Glass | |
34/35 |
Useful for the connection of dissimilar sized ground Glass joints | |
Expansion adapter |