Learn More
Quickfit™ Expansion Socket Adapter
Product Code. 10427820
Click to view available options
Cone Size:
10/19
14/23
19/26
24/29
29/32
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
34/35
40/38
Unit Size:
Each
Pack of 2
Product Code. 10427820
Brand: Quickfit XA52
Only null left
Add to basket
Add to basket
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 19/26
For Use With (Application) Useful for the connection of dissimilar sized ground Glass joints
Material Borosilicate Glass
Product Type Expansion adapter
Standard Taper Joint No. 34/35