Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand:  Quickfit XA43
66.81 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
 View more versions of this product
Product Code. 10881391
  • £163.00 / Pack of 2
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24/29
Borosilicate Glass
29/32
Useful for the connection of dissimilar sized ground Glass joints
Expansion adapter