Learn More
Quickfit™ Glass Stirrer Gland
Introduce stirrer paddles into Quickfit™ flasks and vessels with these glass stirrer glands. Quickfit™ Glass Stirrer Glands provide a vapor-tight seal between paddle and flask.
£168.00 - £813.00
Specifications
For Use With (Equipment) Quickfit™ flasks and vessels
For Use With (Application) For introducing stirrer paddles into Quickfit™ flasks and vessels
Includes Screw cap, Rubber rings and PTFE washers
View More Specs
Products 3
Product Code Brand Standard Taper Joint Price Quantity & Availability  
Product Code Brand Standard Taper Joint Price Quantity & Availability  
10238024
View Documents
Quickfit
ST20/2
19/26
£225.00
Each
In Stock
to see stock.
Add to basket
12003314
View Documents
Quickfit
ST20/3
24/29
£168.00
Each
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
15567990
View Documents
Quickfit
ST20/4
29/32
£813.00
Pack of 5
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
Description
Description
  • Holds a vacuum down to 0.1 torr (13.3Pa)
  • Comprises parallel ground glass sleeve with cup to accept a suitable lubricant
  • Supplied complete with screwcaps rubber rings and PTFE washer
Specifications
Specifications
Quickfit™ flasks and vessels
For introducing stirrer paddles into Quickfit™ flasks and vessels
Screw cap, Rubber rings and PTFE washers