missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Heating Element
Quickfit™ Heating Element is a spare part for use with QSW4 Series Water Still.
Brand: Scilabware QWSH4
377.92 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Chromium plated heating element
- Includes 2 independent re-settable thermostats
- Protect the still in the event of water supply failure

Specifications
50/60 Hz | |
Chromium-plated | |
3 kW | |
Heating Element |
2 Thermostats | |
230 V | |
QWS4 Series Water Still |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quickfit™ Heating Element > Voltage: 230 V
Spot an opportunity for improvement?Share a Content Correction