Learn More
Quickfit™ Metal Clamps for Spherical Joints
Finished with corrosion-resistant nickel for durability. Quickfit™ Metal Clamps for Spherical Joints are designed to secure corresponding Quickfit™ glassware with ball and socket style joints.
Brand:  Quickfit JC13
 View more versions of this product
Product Code. 11322015
  • £81.05 / Pack of 5
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for high temperature work
  • Spring loaded to ensure balanced positive pressure to both the ball and socket joint surface
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5/Pk.
Spherical Joint Clamp
Nickel-plated Metal
Spherical joint size S13