Learn More
Quickfit™ Metal Clamps for Spherical Joints
Finished with corrosion-resistant nickel for durability. Quickfit™ Metal Clamps for Spherical Joints are designed to secure corresponding Quickfit™ glassware with ball and socket style joints.
Brand: Quickfit JC35
This item is not returnable.
View return policy
Description
- Suitable for high temperature work
- Spring loaded to ensure balanced positive pressure to both the ball and socket joint surface
- Available in a variety of sizes
Specifications
2/Pk. | |
Spherical Joint Clamp | |
Nickel-plated Metal | |
Spherical joint size S35 |