Learn More
Scilabware Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
Brand:  Scilabware MA1/11
44.31 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12001603
  • £53.80 / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
•With two parallel necks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14/23
Borosilicate Glass
Multiple neck adapter
14/23