Learn More
Quickfit™ PTFE Screwthread Washers
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
Brand:  Quickfit QW13/6
16.85 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12939283
  • £35.70 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
  • For use with Quickfit™ glass screwthreads
  • Melamine screwcaps with aperture at the top
  • Silicone rubber rings with PTFE washers
  • To determine what components are required: i) Check the size of the glass screw thread (size designation refers to O. D. of thread inmm) ii) Check the O. D. of the sampling tube, thermometer, probe etc. , that is to be passed through the screw cap.
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Washer
PTFE
For Quickfit™ Glass screw threads
13