Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand:  Quickfit DA37
140.27 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 15507800
  • £180.00 / Pack of 5
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cone Size: 45/40
Small socket, large cone
Reduction adapter
Dissimilar sized ground Glass joint
Borosilicate Glass
24/29
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Reduction Socket Adapter > Cone Size: 45/40; Socket Size: 24/29