Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand: Quickfit DA38
175.71 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Specifications
Cone Size: 50/42 | |
Small socket, large cone | |
Reduction adapter |
Dissimilar sized ground Glass joint | |
Borosilicate Glass | |
24/29 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now