Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand: Quickfit DA36
139.47 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Specifications
Cone Size: 40/38 | |
Small socket, large cone | |
Reduction adapter |
Dissimilar sized ground Glass joint | |
Borosilicate Glass | |
24/29 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30