Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
Brand:  Quickfit ST51/18
52.35 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10290219
  • £137.00 / Pack of 2
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
  • With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
  • Ideal for securing thermometers, gas inlet tubes, etc
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14/23
Plastic cap, Silicone Rubber ring and PTFE washer
Cone screw thread adapter
Ideal for securing thermometers, gas inlet tubes
Borosilicate Glass
18