Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
Brand: Quickfit ST52/24
38.06 GBP valid until 2024-06-30
Use promo code "21639" to get your promotional price.
Code : 01
Additional Details : Weight : 0.06120kg
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
- With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
- Ideal for securing thermometers, gas inlet tubes, etc
Specifications
19/26 | |
Plastic cap, Silicone Rubber ring and PTFE washer | |
Cone screw thread adapter |
Ideal for securing thermometers, gas inlet tubes | |
Borosilicate Glass | |
24 |