Learn More
Quickfit™ Sealing Ring for Reaction Vessels
Silicone rubber sheathed with PTFE to provide a chemically resistant and grease free seal
Brand: SVL 296-90
49.04 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.02000kg
Description
- For fitting between the flanges of SVL™ reaction vessels and lids
- Silicone rubber sheathed with PTFE to provide a chemically resistant and grease free seal
Specifications
Reaction vessel | |
Sealing ring | |
1PK |