Learn More
Quickfit™ Sloping Splash Head Adapter
Product Code. p-8003014
Click to view available options
Cone Size:
19/26 (Condenser), 19/26 (Flask)
19/26 (Condenser), 24/29 (Flask)
24/29 (Condenser), 24/29 (Flask)
29/32 (Condenser), 29/32 (Flask)
Unit Size:
Pack of 5
Product Code. 15567970
Brand: Quickfit SH7/0/44
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used in distillation assemblies.
  • Used in distillation assemblies to prevent raw liquid carrying over from flask to condenser
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used in Distillation Assemblies to Prevent Raw liquid “Carrying over” From Flask to Condenser
Packaging Quantity 1 Pack
Cone Size 29/32 (Condenser), 29/32 (Flask)
Product Type Adapter