Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
Brand:  Quickfit MF18/3/SC
38.79 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12048371
  • £94.25 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
  • With sidearm for the attachment of rubber hose etc
  • Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24/29
Borosilicate Glass
19/26
Detachable Plastic screw thread connector
Side-arm tubing adapter