Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
Brand: Quickfit MF18/5/SC
38.79 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Informations supplémentaires : Weight : 0.05076kg
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
- With sidearm for the attachment of rubber hose etc
- Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Spécification
34/35 | |
Borosilicate Glass | |
19/26 |
Detachable Plastic screw thread connector | |
Side-arm tubing adapter |